Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191715.1 | NP_001178644.1 | 1 - 2 | 229 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |