Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025271.1 | NP_001020442.1 | 5 - 6 | 1659 | 64 | |
GenBank mRNA | AF036335.1 | - | 1 - 2 | 188 | 64 | |
BC098731.1 | - | 5 - 6 | 1659 | 7384804 | 64 | |
FQ229853.1 | - | 1 - 2 | 201 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |