Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107914.1 | NP_001101384.1 | 27 - 28 | 4409 | 73 | |
XM_006238934.1 | XP_006238996.1 | 28 - 29 | 4517 | 73 | ||
XM_006238935.2 | XP_006238997.1 | 28 - 29 | 4880 | 73 | ||
XM_006238937.1 | XP_006238999.1 | 27 - 28 | 4418 | 73 | ||
XM_006238938.1 | XP_006239000.1 | 27 - 28 | 4352 | 73 | ||
XM_006238939.1 | XP_006239001.1 | 27 - 28 | 4352 | 73 | ||
XM_008764142.1 | XP_008762364.1 | 26 - 27 | 4253 | 73 | ||
XM_008764143.1 | XP_008762365.1 | 26 - 27 | 4253 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |