Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108682.1 | NP_001102152.1 | 1 | 2972 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |