Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025651.1 | NP_001020822.1 | 5 - 6 | 557 | 78 | |
XM_006239013.2 | XP_006239075.1 | 5 - 6 | 1395 | 78 | ||
GenBank mRNA | BC098679.1 | - | 5 - 6 | 557 | 7384784 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |