Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017284.1 | NP_058980.1 | 4 - 5 | 502 | 60 | |
GenBank mRNA | BC058487.1 | - | 4 - 5 | 522 | 6891616 | 60 |
D21799.1 | - | 4 - 5 | 502 | 60 | ||
FQ217737.1 | - | 4 - 5 | 532 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |