Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017227.1 | NP_058923.1 | 5 - 6 | 542 | 70 | |
XM_006239145.2 | XP_006239207.1 | 5 - 6 | 563 | 70 | ||
GenBank mRNA | AB008803.1 | - | 5 - 6 | 542 | 70 | |
AB010999.1 | - | 5 - 6 | 542 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |