Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134703.1 | NP_001128175.1 | 1 - 2 | 316 | 72 | |
GenBank mRNA | AI070284.1 | - | 1 - 2 | 316 | 72 | |
BF286993.1 | - | 1 - 2 | 309 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |