Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130405.1 | NP_569089.1 | 6 - 7 | 1191 | 70 | |
GenBank mRNA | AF305619.1 | - | 6 - 7 | 1191 | 70 | |
AF393783.1 | - | 6 - 7 | 1108 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |