Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024162.1 | NP_077076.1 | 3 - 4 | 385 | 75 | |
GenBank mRNA | FQ217465.1 | - | 3 - 4 | 389 | 75 | |
J02773.1 | - | 3 - 4 | 385 | 75 | ||
M18034.1 | - | 3 - 4 | 372 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |