Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729579.3 | XP_002729625.1 | 1 - 2 | 133 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |