Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729579.3 | XP_002729625.1 | 4 - 5 | 390 | 103 | |
XM_006239606.2 | XP_006239668.1 | 3 - 4 | 616 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |