Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106692.1 | NP_001100162.1 | 5 - 6 | 540 | 75 | |
XM_008764220.1 | XP_008762442.1 | 5 - 6 | 548 | 75 | ||
GenBank mRNA | FQ231865.1 | - | 5 - 6 | 520 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |