Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013216.2 | NP_001013234.2 | 8 - 9 | 1602 | 67 | |
XM_006239469.2 | XP_006239531.1 | 8 - 9 | 1611 | 67 | ||
XM_008764367.1 | XP_008762589.1 | 8 - 9 | 1640 | 67 | ||
GenBank mRNA | BQ200912.1 | - | 3 - 4 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |