Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031342.1 | NP_112632.1 | 2 - 3 | 160 | 118 | |
GenBank mRNA | AB021645.1 | - | 2 - 3 | 160 | 118 | |
BC070503.1 | - | 2 - 3 | 166 | 7096587 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |