Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199096.1 | NP_954527.1 | 5 - 6 | 510 | 58 | |
XM_006239179.2 | XP_006239241.1 | 5 - 6 | 543 | 58 | ||
GenBank mRNA | BC061826.1 | - | 5 - 6 | 510 | 5599028 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |