Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017209.1 | NP_058905.1 | 8 - 9 | 905 | 66 | |
XM_006239009.2 | XP_006239071.1 | 9 - 10 | 1024 | 66 | ||
GenBank mRNA | AB015724.1 | - | 8 - 9 | 905 | 66 | |
FQ210707.1 | - | 8 - 9 | 901 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |