Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrap73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014262.1 | NP_001014284.1 | 6 - 7 | 623 | 74 | |
GenBank mRNA | BC082062.1 | - | 6 - 7 | 623 | 7132552 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |