Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107979.1 | NP_001101449.1 | 4 - 5 | 577 | 87 | |
XM_008764016.1 | XP_008762238.1 | 4 - 5 | 711 | 87 | ||
XM_008764017.1 | XP_008762239.1 | 4 - 5 | 638 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |