Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239327.2 | XP_006239389.1 | 3 - 4 | 974 | 82 | |
XM_008764305.1 | XP_008762527.1 | 3 - 4 | 974 | 82 | ||
XM_008764306.1 | XP_008762528.1 | 3 - 4 | 974 | 82 | ||
XM_238429.8 | XP_238429.5 | 3 - 4 | 974 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |