Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108689.2 | NP_001102159.1 | 1 - 2 | 219 | 66 | |
XM_006239238.2 | XP_006239300.1 | 1 - 2 | 550 | 66 | ||
XM_006239239.2 | XP_006239301.1 | 3 - 4 | 979 | 66 | ||
XM_006239240.2 | XP_006239302.1 | 3 - 4 | 379 | 66 | ||
GenBank mRNA | BC158607.1 | - | 3 - 4 | 332 | 7192223 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |