Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239238.2 | XP_006239300.1 | 6 - 7 | 1180 | 110 | |
XM_006239239.2 | XP_006239301.1 | 8 - 9 | 1609 | 110 | ||
XM_006239240.2 | XP_006239302.1 | 8 - 9 | 1009 | 110 | ||
XM_006239244.2 | XP_006239306.1 | 8 - 9 | 1013 | 110 | ||
GenBank mRNA | BC158607.1 | - | 8 - 9 | 962 | 7192223 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |