Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239282.2 | XP_006239344.1 | 8 - 9 | 1379 | 68 | |
XM_006239283.2 | XP_006239345.1 | 8 - 9 | 1037 | 68 | ||
XM_006239286.2 | XP_006239348.1 | 7 - 8 | 880 | 68 | ||
XM_008764300.1 | XP_008762522.1 | 9 - 10 | 1367 | 68 | ||
XM_008764301.1 | XP_008762523.1 | 9 - 10 | 1370 | 68 | ||
XM_008764302.1 | XP_008762524.1 | 7 - 8 | 867 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |