Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107994.2 | NP_001101464.1 | 2 - 3 | 1020 | 59 | |
XM_006239462.2 | XP_006239524.1 | 2 - 3 | 1053 | 59 | ||
XM_008764257.1 | XP_008762479.1 | 2 - 3 | 1266 | 59 | ||
XM_008764258.1 | XP_008762480.1 | 1 - 2 | 851 | 59 | ||
XM_008764259.1 | XP_008762481.1 | 2 - 3 | 2997 | 59 | ||
GenBank mRNA | BC158603.1 | - | 2 - 3 | 1020 | 7133644 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |