Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109438.1 | NP_001102908.1 | 15 - 16 | 2409 | 62 | |
XM_006238901.2 | XP_006238963.1 | 14 - 15 | 2485 | 62 | ||
XM_006238902.2 | XP_006238964.1 | 14 - 15 | 2476 | 62 | ||
XM_006238903.2 | XP_006238965.1 | 13 - 14 | 2374 | 62 | ||
XM_006238904.2 | XP_006238966.1 | 13 - 14 | 2365 | 62 | ||
XM_006238906.2 | XP_006238968.1 | 14 - 15 | 2269 | 62 | ||
GenBank mRNA | FQ216284.1 | - | 5 - 6 | 554 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |