Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109267.1 | NP_001102737.1 | 2 - 3 | 211 | 91 | |
XM_006238968.2 | XP_006239030.1 | 2 - 3 | 231 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |