Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109267.1 | NP_001102737.1 | 5 - 6 | 459 | 81 | |
XM_006238968.2 | XP_006239030.1 | 5 - 6 | 479 | 81 | ||
XM_006238969.2 | XP_006239031.1 | 4 - 5 | 415 | 81 | ||
XM_006238970.2 | XP_006239032.1 | 3 - 4 | 294 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |