Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108003.1 | NP_001101473.1 | 2 - 3 | 375 | 70 | |
XM_008764147.1 | XP_008762369.1 | 4 - 5 | 942 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |