Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229519.1 | XP_006229581.1 | 4 - 5 | 560 | 92 | |
XM_215222.7 | XP_215222.5 | 4 - 5 | 696 | 92 | ||
GenBank mRNA | FQ219097.1 | - | 4 - 5 | 454 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |