Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107450.1 | NP_001100920.1 | 6 - 7 | 1166 | 74 | |
XM_008761497.1 | XP_008759719.1 | 6 - 7 | 2318 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |