Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013076.1 | NP_001013094.1 | 4 - 5 | 608 | 103 | |
XM_006233488.1 | XP_006233550.1 | 3 - 4 | 590 | 103 | ||
XM_008761550.1 | XP_008759772.1 | 1 - 2 | 975 | 103 | ||
GenBank mRNA | BC083638.1 | - | 4 - 5 | 608 | 7134832 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |