Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233537.2 | XP_006233599.1 | 9 - 10 | 1376 | 79 | |
XM_227798.8 | XP_227798.5 | 10 - 11 | 2218 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |