Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014177.1 | NP_001014199.1 | 11 - 12 | 1319 | 84 | |
XM_008761517.1 | XP_008759739.1 | 11 - 12 | 1608 | 84 | ||
GenBank mRNA | BC079224.1 | - | 11 - 12 | 1319 | 7132232 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |