Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134549.2 | NP_001128021.2 | 1 - 2 | 1555 | 72 | |
XM_006233489.2 | XP_006233551.1 | 3 - 4 | 1839 | 72 | ||
XM_006233490.2 | XP_006233552.1 | 4 - 5 | 2429 | 72 | ||
XM_006233491.2 | XP_006233553.1 | 3 - 4 | 1839 | 72 | ||
XM_008761551.1 | XP_008759773.1 | 4 - 5 | 1868 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |