Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039005.1 | NP_001034094.1 | 2 - 3 | 286 | 72 | |
GenBank mRNA | BC099787.1 | - | 2 - 3 | 286 | 7934506 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Rattus norvegicus Probe spans exons |