Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033079.1 | NP_001028251.1 | 8 - 9 | 1777 | 97 | |
GenBank mRNA | L06434.2 | - | 8 - 9 | 1777 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |