Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107717.1 | NP_001101187.1 | 6 - 7 | 1418 | 64 | |
XM_006233159.2 | XP_006233221.1 | 7 - 8 | 1426 | 64 | ||
XM_006233160.2 | XP_006233222.1 | 7 - 8 | 1419 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |