Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004227.1 | NP_001004227.1 | 7 - 8 | 748 | 55 | |
XM_008761456.1 | XP_008759678.1 | 6 - 7 | 704 | 55 | ||
GenBank mRNA | BC079374.1 | - | 7 - 8 | 748 | 7110129 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |