Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012083.1 | NP_001012083.1 | 2 - 3 | 339 | 99 | |
XM_006233234.2 | XP_006233296.1 | 2 - 3 | 1934 | 99 | ||
XM_006233235.2 | XP_006233297.1 | 2 - 3 | 1935 | 99 | ||
GenBank mRNA | BC082033.1 | - | 2 - 3 | 339 | 7130739 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |