Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012083.1 | NP_001012083.1 | 5 - 6 | 813 | 162 | |
XM_006233234.2 | XP_006233296.1 | 5 - 6 | 2408 | 162 | ||
XM_006233235.2 | XP_006233297.1 | 5 - 6 | 2409 | 162 | ||
GenBank mRNA | BC082033.1 | - | 5 - 6 | 813 | 7130739 | 162 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |