Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012083.1 | NP_001012083.1 | 7 - 8 | 1101 | 115 | |
XM_006233234.2 | XP_006233296.1 | 7 - 8 | 2696 | 115 | ||
GenBank mRNA | BC082033.1 | - | 7 - 8 | 1101 | 7130739 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |