Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047918.2 | NP_001041383.2 | 9 - 10 | 1585 | 89 | |
GenBank mRNA | FQ231577.1 | - | 8 - 9 | 1265 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |