Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108950.2 | NP_001102420.2 | 5 - 6 | 539 | 60 | |
XM_006233171.1 | XP_006233233.1 | 5 - 6 | 572 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |