Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135879.1 | NP_001129351.1 | 4 - 5 | 397 | 70 | |
XM_006233244.2 | XP_006233306.1 | 6 - 7 | 671 | 70 | ||
XM_006233246.2 | XP_006233308.1 | 6 - 7 | 674 | 70 | ||
XM_006233251.2 | XP_006233313.1 | 6 - 7 | 494 | 70 | ||
XM_006233252.2 | XP_006233314.1 | 5 - 6 | 442 | 70 | ||
XM_006233253.2 | XP_006233315.1 | 5 - 6 | 576 | 70 | ||
XM_006233254.2 | XP_006233316.1 | 5 - 6 | 579 | 70 | ||
XM_008761467.1 | XP_008759689.1 | 5 - 6 | 841 | 70 | ||
XM_008761468.1 | XP_008759690.1 | 5 - 6 | 475 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |