Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014176.2 | NP_001014198.1 | 2 - 3 | 391 | 62 | |
XM_008761466.1 | XP_008759688.1 | 2 - 3 | 421 | 62 | ||
GenBank mRNA | BC079151.1 | - | 2 - 3 | 337 | 7128610 | 62 |
CO400199.1 | - | 2 - 3 | 391 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |