Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199407.1 | NP_955439.1 | 13 - 14 | 2437 | 100 | |
XM_006233402.2 | XP_006233464.1 | 14 - 15 | 2442 | 100 | ||
GenBank mRNA | AB118026.1 | - | 13 - 14 | 2437 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |