Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007640.1 | NP_001007641.1 | 5 - 6 | 811 | 93 | |
XM_006233320.2 | XP_006233382.1 | 6 - 7 | 1941 | 93 | ||
XM_006233321.2 | XP_006233383.1 | 6 - 7 | 927 | 93 | ||
GenBank mRNA | BC079404.1 | - | 5 - 6 | 811 | 7112817 | 93 |
FQ212179.1 | - | 6 - 7 | 902 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |