Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761482.1 | XP_008759704.1 | 8 - 9 | 1403 | 58 | |
XM_008761483.1 | XP_008759705.1 | 7 - 8 | 983 | 58 | ||
XM_227715.7 | XP_227715.5 | 7 - 8 | 1510 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |