Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107729.1 | NP_001101199.1 | 4 - 5 | 898 | 105 | |
XM_006233495.2 | XP_006233557.1 | 4 - 5 | 926 | 105 | ||
XM_006233496.2 | XP_006233558.1 | 4 - 5 | 856 | 105 | ||
XM_006233497.2 | XP_006233559.1 | 4 - 5 | 926 | 105 | ||
XM_006233498.2 | XP_006233560.1 | 4 - 5 | 856 | 105 | ||
GenBank mRNA | BC088420.1 | - | 5 - 6 | 922 | 105 | |
BC158758.1 | - | 4 - 5 | 790 | 7099624 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |