Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107729.1 | NP_001101199.1 | 1 - 2 | 104 | 83 | |
XM_006233495.2 | XP_006233557.1 | 1 - 2 | 132 | 83 | ||
XM_006233497.2 | XP_006233559.1 | 1 - 2 | 132 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |